2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene ≥98.0%
Supplier: TCI America
Synonyms:
3,3'-(2,7-Dibromo-9H-fluorene-9,9-diyl)bis(N,N-dimethylpropan-1-amine)
D4550-1G
D4550-200MG
TCD4550-1GEA
711
CAD
TCD4550-1G
TCD4550-200MG
2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene ≥98.0%
2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene
CAS Number: 673474-73-2
MDL Number: MFCD22988888
Molecular Formula: C23H30Br2N2
Molecular Weight: 494.32
Purity/Analysis Method: >98.0% (HPLC,N)
Melting point (°C): 76
Formula:
C₂₃H₃₀Br₂N₂ MW: 494.32 g/mol Melting Pt: 76 °C |
MDL Number:
MFCD22988888 CAS Number: 673474-73-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...