3,6-Diiodocarbazole ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3,6-Diiodo-9H-carbazole
D4548-25G
D4548-5G
TCD4548-25GEA
570.38
CAD
TCD4548-25G
TCD4548-5G
3,6-Diiodocarbazole ≥98.0% (by GC)
3,6-Diiodocarbazole
CAS Number: 57103-02-3
MDL Number: MFCD00450017
Molecular Formula: C12H7I2N
Molecular Weight: 419.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207
Formula:
C₁₂H₇I₂N MW: 419.00 g/mol Melting Pt: 207 °C |
MDL Number:
MFCD00450017 CAS Number: 57103-02-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...