3,6-Diiodo-9-phenylcarbazole ≥98.0%

Supplier: TCI America

Synonyms: 3,6-Diiodo-9-phenyl-9H-carbazole

D4543-1G D4543-5G
TCD4543-1GEA 63.2 CAD
TCD4543-1G TCD4543-5G
3,6-Diiodo-9-phenylcarbazole ≥98.0%
3,6-Diiodo-9-phenylcarbazole

CAS Number: 57103-21-6
MDL Number: MFCD11521285
Molecular Formula: C18H11I2N
Molecular Weight: 495.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 183

Formula: C₁₈H₁₁I₂N
Melting Pt: 183 °C
MDL Number: MFCD11521285
CAS Number: 57103-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b070788e1234be9bb05023b58d8ae621a820df41&vid=b070788e1234be9bb05023b58d8ae621a820df41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR