Di(trimethylolpropane) ≥98.0% (by GC)

Supplier: TCI America

D4538-25G D4538-500G
TCD4538-25GEA 34.76 CAD
TCD4538-25G TCD4538-500G
Di(trimethylolpropane) ≥98.0% (by GC)
Di(trimethylolpropane)

CAS Number: 23235-61-2
MDL Number: MFCD00192117
Molecular Formula: C12H26O5
Molecular Weight: 250.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 110
Flash Point (°C): 237


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bis[2-ethyl-2,2-bis(hydroxymethyl)ethyl] Ether, 2,2'-Oxybis(methylene)bis(2-ethyl-1,3-propanediol)

Formula: C₁₂H₂₆O₅
Boiling Pt: 215 °C
Melting Pt: 110 °C
Flash Pt: 237 °C
MDL Number: MFCD00192117
CAS Number: 23235-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fed879b254c0d4733e8ae775078094be87fac7e&vid=1fed879b254c0d4733e8ae775078094be87fac7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR