3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Difluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

D4535-25G D4535-5G
TCD4535-25GEA 571.96 CAD
TCD4535-25G TCD4535-5G
3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0%
3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl

CAS Number: 85312-59-0
MDL Number: MFCD09838999
Molecular Formula: C21H24F2
Molecular Weight: 314.42
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₂₁H₂₄F₂
Melting Pt: 68 °C
MDL Number: MFCD09838999
CAS Number: 85312-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cceb4afcfae517e1f515ba8478c5c31b83713942&vid=cceb4afcfae517e1f515ba8478c5c31b83713942&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR