3,4-Difluoro-4'-(trans-4-pentylcyclohexyl)biphenyl ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Difluoro-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl

D4534-25G D4534-5G
TCD4534-25GEA 547.15 CAD
TCD4534-25G TCD4534-5G
3,4-Difluoro-4'-(trans-4-pentylcyclohexyl)biphenyl ≥98.0%
3,4-Difluoro-4'-(trans-4-pentylcyclohexyl)biphenyl

CAS Number: 134412-17-2
MDL Number: MFCD09839000
Molecular Formula: C23H28F2
Molecular Weight: 342.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 58

Formula: C₂₃H₂₈F₂
Melting Pt: 58 °C
MDL Number: MFCD09839000
CAS Number: 134412-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68e116cd8f2ca6584ca70a7be4524ff912776a7c&vid=68e116cd8f2ca6584ca70a7be4524ff912776a7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR