2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: 4-(4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl)benzene-1,3-diol

D4527-25G D4527-5G
TCD4527-25GEA 545.1 CAD
TCD4527-25G TCD4527-5G
2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine ≥98.0%
2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine

CAS Number: 1668-53-7
MDL Number: MFCD16251125
Molecular Formula: C25H23N3O2
Molecular Weight: 397.48
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 205

Formula: C₂₅H₂₃N₃O₂
Melting Pt: 205 °C
MDL Number: MFCD16251125
CAS Number: 1668-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3d18fa5da728ad4f689644dcc2d226e7d9e8acf&vid=f3d18fa5da728ad4f689644dcc2d226e7d9e8acf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR