1,2-Difluoro-4-(methylsulfonyl)benzene ≥98.0% (by GC)

Supplier: TCI America

D4526-1G D4526-5G
TCD4526-1GEA 61.62 CAD
TCD4526-1G TCD4526-5G
1,2-Difluoro-4-(methylsulfonyl)benzene ≥98.0% (by GC)
1,2-Difluoro-4-(methylsulfonyl)benzene

CAS Number: 424792-57-4
MDL Number: MFCD04037934
Molecular Formula: C7H6F2O2S
Molecular Weight: 192.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77

Formula: C7H6F2O2S
Melting Pt: 77 °C
MDL Number: MFCD04037934
CAS Number: 424792-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa835d45cf21f358e6cd741f1acfe64aafd54395&vid=aa835d45cf21f358e6cd741f1acfe64aafd54395&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR