3,6-Dimethylsalicylaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D4507-1G
D4507-5G
TCD4507-1GEA
285.98
CAD
TCD4507-1G
TCD4507-5G
3,6-Dimethylsalicylaldehyde ≥98.0% (by GC, titration analysis)
3,6-Dimethylsalicylaldehyde
CAS Number: 1666-04-2
MDL Number: MFCD04037328
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 61
Other Product Names / Synonyms: 2-Hydroxy-3,6-dimethylbenzaldehyde
Formula:
C₉H₁₀O₂ Melting Pt: 61 °C |
MDL Number:
MFCD04037328 CAS Number: 1666-04-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...