3,5-Dimethyl-4-nitroisoxazole ≥98.0% (by GC)

Supplier: TCI America

D4506-25G D4506-5G
TCD4506-25GEA 271.13 CAD
TCD4506-25G TCD4506-5G
3,5-Dimethyl-4-nitroisoxazole ≥98.0% (by GC)
3,5-Dimethyl-4-nitroisoxazole

CAS Number: 1123-49-5
Molecular Formula: C5H6N2O3
Molecular Weight: 142.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63

Formula: C₅H₆N₂O₃
Melting Pt: 63 °C
MDL Number: MFCD00051656
CAS Number: 1123-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7781f05c006d1ca474bee57dcc8705510fb52ee7&vid=7781f05c006d1ca474bee57dcc8705510fb52ee7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR