2,7-Diiodofluorene ≥98.0% (by GC)

Supplier: TCI America

D4504-1G D4504-5G
TCD4504-1GEA 184.86 CAD
TCD4504-1G TCD4504-5G
2,7-Diiodofluorene ≥98.0% (by GC)
2,7-Diiodofluorene

CAS Number: 16218-28-3
MDL Number: MFCD00040153
Molecular Formula: C13H8I2
Molecular Weight: 418.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 218

Formula: C₁₃H₈I₂
Melting Pt: 218 °C
MDL Number: MFCD00040153
CAS Number: 16218-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f356cc87d9e2cb2aa2b1a631df9be1c3a6cef32&vid=9f356cc87d9e2cb2aa2b1a631df9be1c3a6cef32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR