(R)-(-)-DTBM-SEGPHOS® ≥99.0% (by HPLC)
Supplier: TCI America
Synonyms:
(R)-(-)-4,4'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene), (R)-(-)-DTBM-SEGPHOS®
CAS Number: 566940-03-2
MDL Number: MFCD09753003
Molecular Formula: C74H100O8P2
Molecular Weight: 1179.55
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 128
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (R)-(-)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole, (R)-(-)-4,4'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)
Formula:
C₇₄H₁₀₀O₈P₂ MW: 1179.530 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD09753003 CAS Number: 566940-03-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...