(S)-(+)-DTBM-SEGPHOS® ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-(+)-DTBM-SEGPHOS®, (S)-(+)-4,4'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene), (S)-(+)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole

D4500-1G D4500-200MG
TCD4500-1GEA 228.15 CAD
TCD4500-1G TCD4500-200MG
(S)-(+)-DTBM-SEGPHOS® ≥99.0% (by HPLC)
(S)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole

CAS Number: 210169-40-7
MDL Number: MFCD09753003
Molecular Formula: C74H100O8P2
Molecular Weight: 1179.55
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-(+)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole, (S)-(+)-4,4'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)

Formula: C₇₄H₁₀₀O₈P₂
MW: 1179.530 g/mol
MDL Number: MFCD09753003
CAS Number: 210169-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8cf0688d256972098667d6d7e3c1e04c83f3ad3&vid=a8cf0688d256972098667d6d7e3c1e04c83f3ad3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR