(R)-(+)-DM-SEGPHOS® ≥95.0% (by HPLC)

Supplier: TCI America

D4499-1G D4499-200MG
TCD4499-1GEA 290.88 CAD
TCD4499-1G TCD4499-200MG
(R)-(+)-DM-SEGPHOS® ≥95.0% (by HPLC)
(R)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole

CAS Number: 850253-53-1
Molecular Formula: C46H44O4P2
Molecular Weight: 722.80
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 248
Specific rotation [a]20/D: 114.7 deg (C=0.095, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (R)-(+)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, (R)-(+)-4,4'-Bis[di(3,5-xylyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)

Formula: C₄₆H₄₄O₄P₂
Melting Pt: 248 °C
MDL Number: MFCD09753008
CAS Number: 850253-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=058e741ebce14c53229adb4eb24083bb427033f5&vid=058e741ebce14c53229adb4eb24083bb427033f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR