(S)-(-)-DM-SEGPHOS® ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-(-)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, (S)-(-)-DM-SEGPHOS®, (S)-(-)-4,4'-Bis[di(3,5-xylyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)

D4498-1G D4498-200MG
TCD4498-1GEA 277.76 CAD
TCD4498-1G TCD4498-200MG
(S)-(-)-DM-SEGPHOS® ≥95.0% (by HPLC)
(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole

CAS Number: 210169-57-6
MDL Number: MFCD09753008
Molecular Formula: C46H44O4P2
Molecular Weight: 722.80
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -61 deg (C=0.1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-(-)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, (S)-(-)-4,4'-Bis[di(3,5-xylyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)

Formula: C₄₆H₄₄O₄P₂
MDL Number: MFCD09753008
CAS Number: 210169-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971a7f10f3a9ea7c86f4974834361c061a2a6ba6&vid=971a7f10f3a9ea7c86f4974834361c061a2a6ba6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR