2,7-Dibromo-9-(9-heptadecyl)carbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,7-Dibromo-9-(heptadecan-9-yl)-9H-carbazole

D4490-1G D4490-200MG
TCD4490-1GEA 499.12 CAD
TCD4490-1G TCD4490-200MG
2,7-Dibromo-9-(9-heptadecyl)carbazole ≥98.0% (by HPLC, total nitrogen)
2,7-Dibromo-9-(9-heptadecyl)carbazole

CAS Number: 955964-73-5
MDL Number: MFCD16619379
Molecular Formula: C29H41Br2N
Molecular Weight: 563.46
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 72

Formula: C₂₉H₄₁Br₂N
MW: 563.46 g/mol
Melting Pt: 72 °C
MDL Number: MFCD16619379
CAS Number: 955964-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb22434d1a29f5ac5c8dd4d6781fe049cf42b0db&vid=eb22434d1a29f5ac5c8dd4d6781fe049cf42b0db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR