4,7-Di(2-thienyl)-2,1,3-benzothiadiazole ≥98.0%

Supplier: TCI America

Synonyms: 4,7-Di(thiophen-2-yl)benzo[c][1,2,5]thiadiazole

D4487-1G D4487-200MG
TCD4487-1GEA 820.02 CAD
TCD4487-1G TCD4487-200MG
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole ≥98.0%
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole

CAS Number: 165190-76-1
MDL Number: MFCD18252231
Molecular Formula: C14H8N2S3
Molecular Weight: 300.41
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 124
Lambda max.: 445 nm (CHCl3)

Formula: C₁₄H₈N₂S₃
Melting Pt: 124 °C
MDL Number: MFCD18252231
CAS Number: 165190-76-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd734c1ed4da7549cee79e8fdb7893a459df12fd&vid=fd734c1ed4da7549cee79e8fdb7893a459df12fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR