4,5-Dimethoxy-1-benzocyclobutenecarbonitrile ≥98.0%
Supplier: TCI America
Synonyms:
3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile, 4,5-Dimethoxy-1-cyanobenzocyclobutane, 1-Cyano-4,5-dimethoxybenzocyclobutene
D4485-1G
D4485-5G
TCD4485-1GEA
199.08
CAD
TCD4485-1G
TCD4485-5G
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile ≥98.0%
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile
CAS Number: 35202-54-1
MDL Number: MFCD01846132
Molecular Formula: C11H11NO2
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 86
Other Product Names / Synonyms: 1-Cyano-4,5-dimethoxybenzocyclobutene, 3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile
Formula:
C₁₁H₁₁NO₂ MW: 189.21 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD01846132 CAS Number: 35202-54-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...