1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane ≥98.0% (by GC)

Supplier: TCI America

D4484-25G D4484-500G
TCD4484-25GEA 67.47 CAD
TCD4484-25G TCD4484-500G
1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane ≥98.0% (by GC)
1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane

CAS Number: 132182-92-4
MDL Number: MFCD07784227
Molecular Formula: C7H3F13O
Molecular Weight: 350.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Specific Gravity (20/20): 1.67

Formula: C₇H₃F₁₃O
Boiling Pt: 100 °C
Density: 1.67 (20 °C)
MDL Number: MFCD07784227
CAS Number: 132182-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94ab3c7a97c7b1141402455299b43d8b6ccda4e1&vid=94ab3c7a97c7b1141402455299b43d8b6ccda4e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR