3,6-Dibromo-9-n-octylcarbazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,6-Dibromo-9-octyl-9H-carbazole

D4482-1G D4482-200MG
TCD4482-1GEA 355.34 CAD
TCD4482-1G TCD4482-200MG
3,6-Dibromo-9-n-octylcarbazole ≥98.0% (by GC)
3,6-Dibromo-9-n-octylcarbazole

CAS Number: 79554-93-1
MDL Number: MFCD23703112
Molecular Formula: C20H23Br2N
Molecular Weight: 437.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83

Formula: C₂₀H₂₃Br₂N
MW: 437.22 g/mol
Melting Pt: 83 °C
MDL Number: MFCD23703112
CAS Number: 79554-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9103f611491b23ada5041aa05d43c4c1a8d2b4e2&vid=9103f611491b23ada5041aa05d43c4c1a8d2b4e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR