2,6-Dimethoxyaniline ≥98.0%
Supplier: TCI America
D4480-1G
D4480-5G
TCD4480-1GEA
83.74
CAD
TCD4480-1G
TCD4480-5G
2,6-Dimethoxyaniline ≥98.0%
2,6-Dimethoxyaniline
CAS Number: 2734-70-5
MDL Number: MFCD00053934
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 76
Formula:
C₈H₁₁NO₂ Boiling Pt: 140 °C Melting Pt: 76 °C |
MDL Number:
MFCD00053934 CAS Number: 2734-70-5 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...