4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4478-25G D4478-5G
TCD4478-25GEA 801.69 CAD
TCD4478-25G TCD4478-5G
4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline ≥98.0% (by HPLC, total nitrogen)
4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline

CAS Number: 89114-91-0
MDL Number: MFCD09832972
Molecular Formula: C34H29N
Molecular Weight: 451.61
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Freezing point (°C): 143
Lambda max.: 370 nm (THF)

Formula: C₃₄H₂₉N
MDL Number: MFCD09832972
CAS Number: 89114-91-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d20bb952c4e1a906294c814f5631c72922ecc526&vid=d20bb952c4e1a906294c814f5631c72922ecc526&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR