3,4-Dichlorobenzamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4474-1G D4474-5G
TCD4474-1GEA 49.61 CAD
TCD4474-1G TCD4474-5G
3,4-Dichlorobenzamide ≥98.0% (by HPLC, total nitrogen)
3,4-Dichlorobenzamide

CAS Number: 2670-38-4
MDL Number: MFCD00053009
Molecular Formula: C7H5Cl2NO
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 144

Formula: C₇H₅Cl₂NO
Melting Pt: 144 °C
MDL Number: MFCD00053009
CAS Number: 2670-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=594c8132141a44f011c6d07ff579807ab5b09a16&vid=594c8132141a44f011c6d07ff579807ab5b09a16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR