Dimethoxypillar[5]arene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 12,15,32,35,52,55,72,75,92,95-Decamethoxy-1,3,5,7,9(1,4)-pentabenzenacyclodecaphane, DMpillar[5]arene

D4471-100MG D4471-1G
TCD4471-100MGEA 52.14 CAD
TCD4471-100MG TCD4471-1G
Dimethoxypillar[5]arene ≥98.0% (by HPLC)
Dimethoxypillar[5]arene

CAS Number: 1188423-16-6
Molecular Formula: C45H50O10
Molecular Weight: 750.89
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 249


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: DMpillar[5]arene

Formula: C₄₅H₅₀O₁₀
MW: 750.89 g/mol
Melting Pt: 249 °C
MDL Number: MFCD28386103
CAS Number: 1188423-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2252bc55ca410853d0e137509116ccdb9484e7c2&vid=2252bc55ca410853d0e137509116ccdb9484e7c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR