7-(Diethylamino)coumarin-3-carbonitrile ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 3-Cyano-7-(diethylamino)coumarin, 7-(Diethylamino)-2-oxo-2H-chromene-3-carbonitrile

D4462-1G D4462-5G
TCD4462-1GEA 371.3 CAD
TCD4462-1G TCD4462-5G
7-(Diethylamino)coumarin-3-carbonitrile ≥98.0% (by HPLC, total nitrogen)
7-(Diethylamino)coumarin-3-carbonitrile

CAS Number: 51473-74-6
MDL Number: MFCD00068688
Molecular Formula: C14H14N2O2
Molecular Weight: 242.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 230
Lambda max.: 428 nm (EtOH)


Product Categories: Materials Chemistry, Coumarin Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: 3-Cyano-7-(diethylamino)coumarin

Formula: C₁₄H₁₄N₂O₂
MW: 242.28 g/mol
Melting Pt: 230 °C
MDL Number: MFCD00068688
CAS Number: 51473-74-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8366b2a4147758ea48d594e53a7b2f2371e23c5&vid=d8366b2a4147758ea48d594e53a7b2f2371e23c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR