3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥98.0%

Supplier: TCI America

Synonyms: DOC Iodide, 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzoxazolylidene]-1-propenyl]benzoxazolium Iodide, 3,3'-Diethyloxacarbocyanine iodide, DiOC2(3) iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]oxazol-3-ium iodide

D4453-1G D4453-5G
TCD4453-1GEA 176.06 CAD
TCD4453-1G TCD4453-5G
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥98.0%
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide]

CAS Number: 905-96-4
MDL Number: MFCD00011952
Molecular Formula: C21H21IN2O2
Molecular Weight: 460.32
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 276
Lambda max.: 483 nm (MeOH)


Product Categories: Materials Chemistry, Cyanine & Squarylium Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzoxazolylidene]-1-propenyl]benzoxazolium Iodide, DOC Iodide, DiOC2(3)

Formula: C₂₁H₂₁IN₂O₂
MW: 460.32 g/mol
Melting Pt: 276 °C
MDL Number: MFCD00011952
CAS Number: 905-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35d57bf24ac55119a0a23f783405a8ffbad314e5&vid=35d57bf24ac55119a0a23f783405a8ffbad314e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR