2,4-Dichloroquinoline ≥98.0% (by GC)

Supplier: TCI America

D4452-25G D4452-5G
TCD4452-25GEA 387.1 CAD
TCD4452-25G TCD4452-5G
2,4-Dichloroquinoline ≥98.0% (by GC)
2,4-Dichloroquinoline

CAS Number: 703-61-7
MDL Number: MFCD00023939
Molecular Formula: C9H5Cl2N
Molecular Weight: 198.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 282
Melting point (°C): 65
Lambda max.: 305 nm (MeOH)

Formula: C₉H₅Cl₂N
Boiling Pt: 282 °C
Melting Pt: 65 °C
MDL Number: MFCD00023939
CAS Number: 703-61-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1415feb351ab4132a1a0787ee3c522090665cdf2&vid=1415feb351ab4132a1a0787ee3c522090665cdf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR