4-Decenoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D4449-25ML D4449-5ML
TCD4449-25MLEA 616.2 CAD
TCD4449-25ML TCD4449-5ML
4-Decenoic acid ≥98.0% (by GC, titration analysis)
4-Decenoic acid

CAS Number: 26303-90-2
MDL Number: MFCD03092963
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.92

Formula: C₁₀H₁₈O₂
Density: 0.92 (20 °C)
MDL Number: MFCD03092963
CAS Number: 26303-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a91e6abc2c0d57c27bb7a44b5c5438956293e041&vid=a91e6abc2c0d57c27bb7a44b5c5438956293e041&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR