4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4,7-Dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine

D4442-1G D4442-200MG
TCD4442-1GEA 385.2 CAD
TCD4442-1G TCD4442-200MG
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine ≥97.0% (by GC)
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine

CAS Number: 333432-27-2
MDL Number: MFCD23703147
Molecular Formula: C5HBr2N3S
Molecular Weight: 294.95
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 148
Storage Temperature: 0-10°C

Formula: C₅HBr₂N₃S
MW: 294.95 g/mol
Melting Pt: 148 °C
MDL Number: MFCD23703147
CAS Number: 333432-27-2

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR