4,4'-Dichlorobenzil ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Dichlorodibenzoyl, 4,4'-Dichlorobibenzoyl, 1,2-Bis(4-chlorophenyl)ethane-1,2-dione
D4440-1G
D4440-5G
TCD4440-1GEA
143.78
CAD
TCD4440-1G
TCD4440-5G
4,4'-Dichlorobenzil ≥98.0%
4,4'-Dichlorobenzil
CAS Number: 3457-46-3
MDL Number: MFCD00018685
Molecular Formula: C14H8Cl2O2
Molecular Weight: 279.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 199
Lambda max.: 272 nm (EtOH)
Other Product Names / Synonyms: 4,4'-Dichlorobibenzoyl, 4,4'-Dichlorodibenzoyl
Formula:
C₁₄H₈Cl₂O₂ MW: 279.12 g/mol Boiling Pt: 220 °C Melting Pt: 199 °C |
MDL Number:
MFCD00018685 CAS Number: 3457-46-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...