Diethyl (3-bromopropyl)phosphonate ≥95.0% (by GC)
Supplier: TCI America
D4434-1G
D4434-5G
TCD4434-1GEA
53.02
CAD
TCD4434-1G
TCD4434-5G
Diethyl (3-bromopropyl)phosphonate ≥95.0% (by GC)
Diethyl (3-bromopropyl)phosphonate
CAS Number: 1186-10-3
MDL Number: MFCD00800966
Molecular Formula: C7H16BrO3P
Molecular Weight: 259.08
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Specific Gravity (20/20): 1.34
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (3-Bromopropyl)phosphonic Acid Diethyl Ester
Formula:
C₇H₁₆BrO₃P Boiling Pt: 97 °C Density: 1.34 (20 °C) |
MDL Number:
MFCD00800966 CAS Number: 1186-10-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...