3,6-Diphenylcarbazole ≥99.0%

Supplier: TCI America

Synonyms: 3,6-Diphenyl-9H-carbazole

D4433-1G D4433-5G
TCD4433-1GEA 66.36 CAD
TCD4433-1G TCD4433-5G
3,6-Diphenylcarbazole ≥99.0%
3,6-Diphenylcarbazole

CAS Number: 56525-79-2
MDL Number: MFCD00222619
Molecular Formula: C24H17N
Molecular Weight: 319.41
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 184

Formula: C₂₄H₁₇N
Melting Pt: 184 °C
MDL Number: MFCD00222619
CAS Number: 56525-79-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54622e7ead1b96f9fc26d7727108595a551cb53c&vid=54622e7ead1b96f9fc26d7727108595a551cb53c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR