4-(N,N-Dimethylamino)phenylboronic acid 97.0-113.0% (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-(Dimethylamino)benzeneboronic acid
D4428-1G
D4428-5G
TCD4428-1GEA
189.54
CAD
TCD4428-1G
TCD4428-5G
4-(N,N-Dimethylamino)phenylboronic acid 97.0-113.0% (contains varying amounts of Anhydride)
4-(N,N-Dimethylamino)phenylboronic acid
CAS Number: 28611-39-4
MDL Number: MFCD01074642
Molecular Formula: C8H12BNO2
Molecular Weight: 165.00
Form: Crystal
Color: White
Melting point (°C): 227
Other Product Names / Synonyms: 4-(Dimethylamino)benzeneboronic Acid
Formula:
C₈H₁₂BNO₂ Melting Pt: 227 °C |
MDL Number:
MFCD01074642 CAS Number: 28611-39-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...