3,5-Dimethyl-4-nitropyrazole ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dimethyl-4-nitro-1H-pyrazole

D4425-25G D4425-5G
TCD4425-25GEA 320.74 CAD
TCD4425-25G TCD4425-5G
3,5-Dimethyl-4-nitropyrazole ≥98.0%
3,5-Dimethyl-4-nitropyrazole

CAS Number: 14531-55-6
MDL Number: MFCD00052513
Molecular Formula: C5H7N3O2
Molecular Weight: 141.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 325
Melting point (°C): 127
Lambda max.: 274 nm (CHCl3)

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
Boiling Pt: 325 °C
Melting Pt: 127 °C
MDL Number: MFCD00052513
CAS Number: 14531-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59d90031664bebb2171b047acc74d85f7b0676bb&vid=59d90031664bebb2171b047acc74d85f7b0676bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR