2,6-Dichloropyridine-4-methanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine

D4423-1G D4423-5G
TCD4423-1GEA 89.27 CAD
TCD4423-1G TCD4423-5G
2,6-Dichloropyridine-4-methanol ≥98.0% (by GC)
2,6-Dichloropyridine-4-methanol

CAS Number: 101990-69-6
MDL Number: MFCD00052638
Molecular Formula: C6H5Cl2NO
Molecular Weight: 178.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136


Other Product Names / Synonyms: 2,6-Dichloro-4-(hydroxymethyl)pyridine

Formula: C₆H₅Cl₂NO
MW: 178.0160 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00052638
CAS Number: 101990-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31cb1f78d84b898b095ba0a2f95e5407f45e2835&vid=31cb1f78d84b898b095ba0a2f95e5407f45e2835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR