2,2-Dimethylsuccinic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2-Dimethylsuccinicanhydride
D4417-25G
D4417-5G
TCD4417-25GEA
254.54
CAD
TCD4417-25G
TCD4417-5G
2,2-Dimethylsuccinic anhydride ≥98.0% (by GC, titration analysis)
2,2-Dimethylsuccinic anhydride
CAS Number: 17347-61-4
MDL Number: MFCD00022608
Molecular Formula: C6H8O3
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 30
Flash Point (°C): 108
Formula:
C₆H₈O₃ MW: 128.1259 g/mol Boiling Pt: 220 °C Melting Pt: 30 °C Flash Pt: 108 °C |
MDL Number:
MFCD00022608 CAS Number: 17347-61-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...