2,3-Dichloro-5-nitropyridine ≥98.0% (by GC)

Supplier: TCI America

D4416-1G D4416-5G
TCD4416-1GEA 47.4 CAD
TCD4416-1G TCD4416-5G
2,3-Dichloro-5-nitropyridine ≥98.0% (by GC)
2,3-Dichloro-5-nitropyridine

CAS Number: 22353-40-8
MDL Number: MFCD03840432
Molecular Formula: C5H2Cl2N2O2
Molecular Weight: 192.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 53
Flash Point (°C): 110

Formula: C₅H₂Cl₂N₂O₂
Boiling Pt: 256 °C
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD03840432
CAS Number: 22353-40-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac913fbb564bd18783cd1c48d7946050c75f04e6&vid=ac913fbb564bd18783cd1c48d7946050c75f04e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR