3,6-Dibromo-2-picoline ≥98.0%

Supplier: TCI America

Synonyms: 3,6-Dibromo-2-methylpyridine, 2,5-Dibromo-6-methylpyridine

D4415-5G D4415-1G
TCD4415-5GEA 85.32 CAD
TCD4415-5G TCD4415-1G
3,6-Dibromo-2-picoline ≥98.0%
3,6-Dibromo-2-picoline

CAS Number: 39919-65-8
MDL Number: MFCD06254589
Molecular Formula: C6H5Br2N
Molecular Weight: 250.92
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 38


Other Product Names / Synonyms: 3,6-Dibromo-2-picoline

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 38 °C
MDL Number: MFCD06254589
CAS Number: 39919-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f8351747bc7a6a4b4eac3d97af14c550733bcec&vid=4f8351747bc7a6a4b4eac3d97af14c550733bcec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR