Dimethyl-7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate ≥95.0%
Supplier: TCI America
Synonyms:
7-Oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylic Acid Dimethyl Ester, Dimethyl 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylate, 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylic Acid Dimethyl Ester
CAS Number: 1829-60-3
Molecular Formula: C10H10O5
Molecular Weight: 210.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Specific Gravity (20/20): 1.27
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 7-Oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylic Acid Dimethyl Ester, Dimethyl 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylate, 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylic Acid Dimethyl Ester
Formula:
C₁₀H₁₀O₅ MW: 210.19 g/mol Boiling Pt: 120 °C Density: 1.27 (20 °C) |
CAS Number:
1829-60-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...