N,N'-Dihydroxypyromellitimide ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4413-1G D4413-5G
TCD4413-1GEA 243.32 CAD
TCD4413-1G TCD4413-5G
N,N'-Dihydroxypyromellitimide ≥96.0% (by HPLC, total nitrogen)
N,N'-Dihydroxypyromellitimide

CAS Number: 57583-53-6
Molecular Formula: C10H4N2O6
Molecular Weight: 248.15
Purity/Analysis Method: >96.0% (HPLC,N)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: N,N'-Dihydroxypyromellitic Diimide, NDHPI

Formula: C₁₀H₄N₂O₆
MDL Number: MFCD28334210
CAS Number: 57583-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ba2589ab271a13a5b054f1bca7f315fb7d4b762&vid=6ba2589ab271a13a5b054f1bca7f315fb7d4b762&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR