9,10-Dihydro-9,10-[3,4]thiophenoanthracene ≥98.0% (by GC)

Supplier: TCI America

D4412-1G D4412-200MG
TCD4412-1GEA 1048.01 CAD
TCD4412-1G TCD4412-200MG
9,10-Dihydro-9,10-[3,4]thiophenoanthracene ≥98.0% (by GC)
9,10-Dihydro-9,10-[3,4]thiophenoanthracene

CAS Number: 42490-26-6
Molecular Formula: C18H12S
Molecular Weight: 260.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 268

Formula: C₁₈H₁₂S
Melting Pt: 268 °C
CAS Number: 42490-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=529dc1c1075479b86fc3f13ff75030dbdc2f7b4d&vid=529dc1c1075479b86fc3f13ff75030dbdc2f7b4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR