2-(Diphenylphosphino)benzaldehyde ≥97.0%

Supplier: TCI America

Synonyms: (2-Formylphenyl)diphenylphosphine

D4411-1G D4411-5G
TCD4411-1GEA 66.36 CAD
TCD4411-1G TCD4411-5G
2-(Diphenylphosphino)benzaldehyde ≥97.0%
2-(Diphenylphosphino)benzaldehyde

CAS Number: 50777-76-9
MDL Number: MFCD00013367
Molecular Formula: C19H15OP
Molecular Weight: 290.30
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 117
Lambda max.: 376 nm (CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: (2-Formylphenyl)diphenylphosphine

Formula: C₁₉H₁₅OP
MW: 290.30 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00013367
CAS Number: 50777-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab11b1fe0fff4242c3665eb6e1e13586f1e706be&vid=ab11b1fe0fff4242c3665eb6e1e13586f1e706be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR