2,2-Dichlorocyclopropylbenzene ≥98.0% (by GC)

Supplier: TCI America

D4410-25G D4410-5G
TCD4410-25GEA 115.66 CAD
TCD4410-25G TCD4410-5G
2,2-Dichlorocyclopropylbenzene ≥98.0% (by GC)
2,2-Dichlorocyclopropylbenzene

CAS Number: 2415-80-7
MDL Number: MFCD00001272
Molecular Formula: C9H8Cl2
Molecular Weight: 187.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 104
Specific Gravity (20/20): 1.23
Specific rotation [a]20/D: 3.18 deg


Other Product Names / Synonyms: 1,1-Dichloro-2-phenylcyclopropane

Formula: C₉H₈Cl₂
Boiling Pt: 105 °C
Density: 1.23 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00001272
CAS Number: 2415-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef33aa8f2d583d8ec7258fb66ed4ca1406dbccef&vid=ef33aa8f2d583d8ec7258fb66ed4ca1406dbccef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR