N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D4404-1G D4404-200MG
TCD4404-1GEA 541.94 CAD
TCD4404-1G TCD4404-200MG
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate ≥98.0% (by titrimetric analysis)
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate

CAS Number: 118612-00-3
MDL Number: MFCD01074420
Molecular Formula: C32H12BF20N
Molecular Weight: 801.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 227

Formula: C₃₂H₁₂BF₂₀N
Melting Pt: 227 °C
MDL Number: MFCD01074420
CAS Number: 118612-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b528c40be4a9bc0695e570cf33bd667ed9207841&vid=b528c40be4a9bc0695e570cf33bd667ed9207841&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR