Dimethyl-1,4-naphthalenedicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Dimethyl naphthalene-1,4-dicarboxylate, 1,4-Naphthalenedicarboxylic Acid Dimethyl Ester
D4400-1G
D4400-5G
TCD4400-1GEA
352.02
CAD
TCD4400-1G
TCD4400-5G
Dimethyl-1,4-naphthalenedicarboxylate ≥98.0%
Dimethyl-1,4-naphthalenedicarboxylate
CAS Number: 7487-15-2
MDL Number: MFCD04038075
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 161
Melting point (°C): 67
Other Product Names / Synonyms: 1,4-Naphthalenedicarboxylic Acid Dimethyl Ester
Formula:
C₁₄H₁₂O₄ Boiling Pt: 161 °C Melting Pt: 67 °C |
MDL Number:
MFCD04038075 CAS Number: 7487-15-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...