6,7-Dimethoxy-4-methylcoumarin ≥98.0%

Supplier: TCI America

D4390-1G D4390-5G
TCD4390-1GEA 48.98 CAD
TCD4390-1G TCD4390-5G
6,7-Dimethoxy-4-methylcoumarin ≥98.0%
6,7-Dimethoxy-4-methylcoumarin

CAS Number: 4281-40-7
MDL Number: MFCD00012338
Molecular Formula: C12H12O4
Molecular Weight: 220.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 138
Lambda max.: 340 nm (MeOH)

Formula: C₁₂H₁₂O₄
MW: 220.22 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00012338
CAS Number: 4281-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0188fc0530aaa6e943742bfc8f8b29981a1c315&vid=b0188fc0530aaa6e943742bfc8f8b29981a1c315&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR