4',7-Dimethoxyisoflavone ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
7-Methoxy-3-(4-methoxyphenyl)chromen-4-one, 7-Methoxy-3-(4-methoxyphenyl)-4-benzopyrone, 4',7-Dimethoxyisoflavone
D4389-1G
D4389-5G
TCD4389-1GEA
155.31
CAD
TCD4389-1G
TCD4389-5G
4',7-Dimethoxyisoflavone ≥97.0% (by GC)
4',7-Dimethoxyisoflavone
CAS Number: 1157-39-7
MDL Number: MFCD00075889
Molecular Formula: C17H14O4
Molecular Weight: 282.30
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 163
Lambda max.: 260 nm (MeOH)
Other Product Names / Synonyms: 7-Methoxy-3-(4-methoxyphenyl)chromone
Formula:
C₁₇H₁₄O₄ MW: 282.3 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00075889 CAS Number: 1157-39-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...