2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) ≥95.0%
Supplier: TCI America
Synonyms:
2,6-Di-tert-butyl-4-methylcyclohexanone
D4385-1ML
D4385-5ML
TCD4385-1MLEA
112.34
CAD
TCD4385-1ML
TCD4385-5ML
2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) ≥95.0%
2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers)
CAS Number: 23790-39-8
Molecular Formula: C15H28O
Molecular Weight: 224.39
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Specific Gravity (20/20): 0.89
Formula:
C₁₅H₂₈O MW: 224.39 g/mol Boiling Pt: 134 °C Density: 0.89 (20 °C) |
MDL Number:
MFCD27976821 CAS Number: 23790-39-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...