2,3-Dibromo-5-chloropyridine ≥98.0% (by GC)

Supplier: TCI America

D4381-25G D4381-5G
TCD4381-25GEA 230.68 CAD
TCD4381-25G TCD4381-5G
2,3-Dibromo-5-chloropyridine ≥98.0% (by GC)
2,3-Dibromo-5-chloropyridine

CAS Number: 137628-17-2
MDL Number: MFCD08687254
Molecular Formula: C5H2Br2ClN
Molecular Weight: 271.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 43

Formula: C₅H₂Br₂ClN
MW: 271.3400 g/mol
Melting Pt: 43 °C
MDL Number: MFCD08687254
CAS Number: 137628-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7d0c1d23e64ec94fdc0913c8da3e227c22578cd&vid=c7d0c1d23e64ec94fdc0913c8da3e227c22578cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR