4,6-Dimethylisophthalic acid ≥95.0%
Supplier: TCI America
D4377-1G
D4377-200MG
TCD4377-1GEA
630.42
CAD
TCD4377-1G
TCD4377-200MG
4,6-Dimethylisophthalic acid ≥95.0%
4,6-Dimethylisophthalic acid
CAS Number: 2790-09-2
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 298
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Melting Pt: 298 °C |
MDL Number:
MFCD00230851 CAS Number: 2790-09-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...